Isothiazole 98%

Supplier: Ambeed
Danger

A669391-250MG A669391-5G
103840-490EA 212 USD
103840-490 76806-526 77308-084
Isothiazole 98%
Isothiazole

Precaución: for Research Use Only

Formula: C₃H₃NS
Storage Temperature: Freezer
MDL Number: MFCD00020818
CAS Number: 288-16-4
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR