Isothiazolin-3-one 95%

Supplier: Ambeed
Warning

A510573-1G A510573-100MG A510573-250MG A510573-5G
103828-384EA 28 USD
103828-384 76843-474 77034-590 77034-592 77034-594
Isothiazolin-3-one 95%
Isothiazolin-3-one

Precaución: for Research Use Only

Formula: C₃H₃NOS
MW: 101.13 g/mol
Melting Pt: 73…74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834764
CAS Number: 1003-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR