Isoquinoline-6-carbonitrile 97%

Supplier: Ambeed
Warning

A220632-250MG A220632-5G A220632-1G
103818-866EA 67 USD
103818-866 103849-748 76770-802 77088-342 77303-558
Isoquinoline-6-carbonitrile 97%
Isoquinoline-6-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₆N₂
MW: 154.1680 g/mol
Boiling Pt: 341.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09991660
CAS Number: 106778-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR