Isoquinoline-5-sulfonyl chloride hydrochloride 95%
Supplier: Ambeed
|
Danger
|
A456923-250MG
A456923-1G
A456923-5G
103827-864EA
33
USD
103827-864
103838-940
76804-584
76842-800
76842-802
Isoquinoline-5-sulfonyl chloride hydrochloride 95%
Isoquinoline-5-sulfonyl chloride hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₇Cl₂NO₂S Melting Pt: 173…183 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08272853 CAS Number: 105627-79-0 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...