5-Isoquinolinecarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isoquinoline-5-carboxylic acid
A113929-250MG
A113929-1G
A113929-5G
A113929-10G
103798-322EA
24
USD
103798-322
103824-164
76796-968
77310-984
77310-986
77623-952
5-Isoquinolinecarboxylic acid 98%
5-Isoquinolinecarboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Boiling Pt: 405.6 °C (760 mm Hg) Melting Pt: 278…281 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03788744 CAS Number: 27810-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...