4-Isoquinolinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isoquinoline-4-carboxylic acid

A194126-1G A194126-25G A194126-5G A194126-250MG A194126-10G
103816-620EA 32 USD
103816-620 103847-230 77085-140 77085-142 77085-144 77164-354 77683-518
4-Isoquinolinecarboxylic acid 98%
4-Isoquinolinecarboxylic acid

Precaución: For research use only.

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Boiling Pt: 405.6 °C (760 mm Hg)
Melting Pt: 265…275 °C
Flash Pt: 199.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00094344
CAS Number: 7159-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR