Isoquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A178875-10G A178875-250MG A178875-5G
103807-380EA 18 USD
103807-380 103845-720 76799-660 76799-662 77301-584
Isoquinoline-3-carboxylic acid 97%
Isoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Boiling Pt: 366.4 °C (760 mm Hg)
Melting Pt: 166…167 °C
Flash Pt: 175.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075137
CAS Number: 6624-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR