Isoquinoline-8-amine 98%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-8-amine, 8-Aminoisoquinoline

A151074-250MG A151074-1G A151074-5G
103804-228EA 14 USD
103804-228 103842-374 77162-804 77317-914 77317-916
Isoquinoline-8-amine 98%
Isoquinoline-8-amine

Precaución: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₉H₈N₂
MW: 144.1700 g/mol
Boiling Pt: 289.8 °C (760 mm Hg)
Melting Pt: 155…157 °C
Flash Pt: 183.349 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00179553
CAS Number: 23687-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR