6-Isoquinolinol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Hydroxyisoquinoline, Isoquinolin-6-ol
A216476-1G
A216476-5G
A216476-100G
A216476-100MG
A216476-10G
103818-560EA
20
USD
103818-560
103849-280
77087-866
77087-868
77165-106
77165-108
77165-110
6-Isoquinolinol 98%
6-Isoquinolinol
Precaución: for Research Use Only
Formula:
C₉H₇NO MW: 145.16 g/mol Boiling Pt: 332.1 °C (760 mm Hg) Melting Pt: 220 °C Flash Pt: 154.6 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114860 CAS Number: 7651-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...