5-Isoquinolineboronic acid 97%

Supplier: Ambeed
Warning

A193681-25G A193681-5G A193681-10G A193681-250MG A193681-1G
103816-562EA 20 USD
103816-562 103847-176 77085-072 77085-074 77260-576 77260-578 77302-348
5-Isoquinolineboronic acid 97%
5-Isoquinolineboronic acid

Precaución: for Research Use Only

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Boiling Pt: 419.1 °C (760 mm Hg)
Melting Pt: 166…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03839356
CAS Number: 371766-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR