Isoquinoline-5-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Isoquinolin-5-amine, 5-Aminoisoquinoline
A100451-10G
A100451-25G
A100451-500G
A100451-5G
A100451-100G
103795-446EA
6
USD
103795-446
103821-142
76686-990
76686-992
76686-994
77296-692
77623-808
Isoquinoline-5-amine 98%
Isoquinoline-5-amine
Precaución: for Research Use Only
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 125…128 °C Flash Pt: 183.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006907 CAS Number: 1125-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...