4-Isoquinolinol 95%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-4-ol, 4-Hydroxyisoquinoline

A297984-100MG A297984-10G A297984-250MG A297984-1G A297984-25G A297984-5G
103814-168EA 25 USD
103814-168 103835-312 76802-504 76802-506 76802-508 76838-002 76838-004 76838-006
4-Isoquinolinol 95%
4-Isoquinolinol

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.1580 g/mol
Boiling Pt: 425.4 °C (760 mm Hg)
Melting Pt: 223…225 °C
Flash Pt: 211.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00234514
CAS Number: 3336-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR