Isoquinoline-4-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminoisoquinoline, Isoquinolin-4-amine
A132391-100G
A132391-10G
A132391-250MG
A132391-1G
A132391-5G
103801-732EA
14
USD
103801-732
76688-688
76688-690
76688-692
77314-310
77314-312
Isoquinoline-4-amine 98%
Isoquinoline-4-amine
Precaución: for Research Use Only
Formula:
C₉H₈N₂ MW: 144.1700 g/mol Boiling Pt: 360 °C (760 mm Hg) Melting Pt: 107…113 °C Flash Pt: 198.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034752 CAS Number: 23687-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...