Isoquinoline-3-amine 97%

Supplier: Ambeed
Warning

Synonyms: Isoquinolin-3-amine, 3-Aminoisoquinoline

A210781-1G A210781-5G A210781-250MG
103818-040EA 12 USD
103818-040 103848-732 77087-116 77087-118 77748-313
Isoquinoline-3-amine 97%
Isoquinoline-3-amine

Precaución: for Research Use Only

Formula: C₉H₈N₂
MW: 144.1700 g/mol
Boiling Pt: 331.2 °C (760 mm Hg)
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00102190
CAS Number: 25475-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR