Isopropyltriphenylphosphonium iodide 97%

Supplier: Ambeed
Warning

A116662-100G A116662-10G A116662-25G A116662-500G A116662-5G
103824-646EA 9 USD
103824-646 76797-090 76797-092 76797-094 76797-096 77297-490
Isopropyltriphenylphosphonium iodide 97%
Isopropyltriphenylphosphonium iodide

Precaución: for Research Use Only

Formula: C₂₁H₂₂IP
Melting Pt: 194…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00031595
CAS Number: 24470-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR