IPTG (isopropyl-β-D-thiogalactopyranoside) 98%

Supplier: Ambeed
Warning

A193402-10G A193402-25G A193402-5G A193402-100G A193402-100MG
103847-150EA 14 USD
103847-150 76800-062 76800-064 77085-052 77085-054 77550-246 77879-228
IPTG (isopropyl-β-D-thiogalactopyranoside) 98%
IPTG (isopropyl-β-D-thiogalactopyranoside)

Precaución: for Research Use Only

Formula: C₉H₁₈O₅S
Boiling Pt: 438.4 °C (760 mm Hg)
Melting Pt: 105 °C
Flash Pt: 219 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR