Isopropyl benzenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Benzenesulphonic acid isopropyl ester

A250426-1G A250426-5G A250426-10G A250426-25G
103811-924EA 5 USD
103811-924 103851-764 77091-116 77091-118 77165-956 77304-528
Isopropyl benzenesulfonate 97%
Isopropyl benzenesulfonate

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
Boiling Pt: 291.7 °C (760 mm Hg)
Flash Pt: 130.2 °C
Storage Temperature: Ambient
MDL Number: MFCD04038771
CAS Number: 6214-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR