Isopropyl-3-aminobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Isopropyl 3-aminobenzoate

A100373-10G A100373-1G
103795-402EA 9 USD
103795-402 103821-100 77008-266 77296-656
Isopropyl-3-aminobenzoate 95%
Isopropyl-3-aminobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.2200 g/mol
Boiling Pt: 301.2 °C (760 mm Hg)
Flash Pt: 156.5 °C
Storage Temperature: Ambient
MDL Number: MFCD07601158
CAS Number: 35005-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR