Isonicotinamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Pyridine-4-carboxamidine hydrochloride, Isonicotinimidamide hydrochloride, 4-Carbamimidoylpyridine hydrochloride, 4-Amidinopyridine hydrochloride, Pyridine-4-carboximidamide hydrochloride, 4-Amidinopyridinium chloride

A125057-100G A125057-10G A125057-250MG A125057-25G A125057-500G A125057-1G A125057-5G
103800-614EA 9 USD
103800-614 77108-800 77108-802 77108-804 77108-806 77108-808 77312-764 77312-766
Isonicotinamidine hydrochloride 97%
Isonicotinamidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇N₃·ClH
MW: 157.601 g/mol
Boiling Pt: 240.7 °C (760 mm Hg)
Melting Pt: 240…242 °C
Flash Pt: 99.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040385
CAS Number: 6345-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR