Isoindolin-1-one 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-1H-isoindol-1-one, 1-Isoindolinone

A147533-100G A147533-250MG A147533-1G A147533-5G A147533-10G
103803-796EA 6 USD
103803-796 103841-900 76798-492 76798-494 77317-276 77317-278 77567-044
Isoindolin-1-one 98%
Isoindolin-1-one

Precaución: For research use only.

Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 380.6 °C (760 mm Hg)
Flash Pt: 220.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03085939
CAS Number: 480-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR