Iron(III) trifluoromethanesulphonate 98%
Supplier: Ambeed
|
Warning
|
A151954-100G
A151954-10G
A151954-1G
A151954-25G
103804-324EA
6
USD
103804-324
103842-474
77028-174
77028-176
77318-058
77318-060
77318-062
Iron(III) trifluoromethanesulphonate 98%
Iron(III) trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₃F₉FeO₉S₃ MW: 503.06 g/mol Boiling Pt: 162 °C (760 mm Hg) Melting Pt: 183 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD15144785 CAS Number: 63295-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...