Iron(II) trifluoromethanesulphonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Iron (II) Trifluoromethanesulphonate
A308048-1G
A308048-5G
A308048-100G
A308048-250MG
A308048-25G
A308048-10G
103814-544EA
6
USD
103814-544
103835-720
76838-476
76838-478
77032-554
77032-556
77305-976
77672-564
Iron(II) trifluoromethanesulphonate 98%
Iron(II) trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₂F₆FeO₆S₂ MW: 353.99 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD08460524 CAS Number: 59163-91-6 UN: 3260 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...