Iron(II) acetylacetonate 95%

Supplier: Ambeed
Warning

A118234-100G A118234-10G A118234-25G
103824-900EA 6 USD
103824-900 76797-216 76797-218 76797-220
Iron(II) acetylacetonate 95%
Iron(II) acetylacetonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄FeO₄
MW: 254.07 g/mol
Boiling Pt: 187.6 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 71.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00144506
CAS Number: 14024-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR