(S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate 98%

Supplier: Ambeed
Danger

Synonyms: (+)-Irinotecan, CPT 11

A164809-250MG A164809-100MG
103805-908EA 164 USD
103805-908 103844-164 77567-402 77615-350
(S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate 98%
(S)-4,11-Diethyl-4-hydroxy-3,14-dioxo-3,4,12,14-tetrahydro-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinolin-9-yl [1,4'-bipiperidine]-1'-carboxylate

Precaución: For research use only. Sealed in dry, at 2…8°C

Formula: C₃₃H₃₈N₄O₆
MW: 586.67 g/mol
Melting Pt: 222…223 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00866307
CAS Number: 97682-44-5
UN: 1544
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR