Iridium(III) chloride hydrate 98%

Supplier: Ambeed
Warning

Synonyms: Iridium trichloride hydrate, Iridium(III) chloride xhydrate

A446060-100MG A446060-250MG A446060-1G A446060-5G
103827-734EA 209 USD
103827-734 103838-798 76804-442 76804-444 77307-342 77307-344
Iridium(III) chloride hydrate 98%
Iridium(III) chloride hydrate

Precaución: for Research Use Only

Formula: Cl₃Ir·xH₂O
MW: 298.58 g/mol (anhy)
Melting Pt: 763 °C
Storage Temperature: Ambient
MDL Number: MFCD00149750
CAS Number: 14996-61-3
Merck Index: 14,05088

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR