Iridium(III) chloride trihydrate 98% I
Supplier: Ambeed
|
Warning
|
A809615-100MG
A809615-250MG
A809615-1G
A809615-5G
103841-086EA
1214
USD
103841-086
76807-566
76807-568
77308-362
77308-364
Iridium(III) chloride trihydrate 98% I
Iridium(III) chloride trihydrate
Precaución: for Research Use Only
Formula:
Cl₃Ir·₃H₂O Melting Pt: 763 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149750 CAS Number: 13569-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...