Iridium(III) chloride 99.9%
Supplier: Ambeed
|
Warning
|
A513338-100MG
A513338-250MG
A513338-1G
A513338-5G
103828-428EA
275
USD
103828-428
103839-528
76805-072
76805-074
77307-670
77307-672
Iridium(III) chloride 99.9%
Iridium(III) chloride
Precaución: for Research Use Only
Formula:
Cl₃Ir Melting Pt: 763 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011063 CAS Number: 10025-83-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...