Indoline-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A139654-100MG A139654-250MG A139654-5G
103802-758EA 46 USD
103802-758 77010-312 77010-314 77315-854
Indoline-5-carboxylic acid 97%
Indoline-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.17 g/mol
Boiling Pt: 369 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07371642
CAS Number: 15861-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR