Indole-7-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-7-carboxylic acid

A273640-250MG A273640-1G A273640-25G A273640-5G A273640-10G
103813-150EA 10 USD
103813-150 103853-072 76692-886 77092-874 77092-876 77092-878 77601-202
Indole-7-carboxylic acid 98%
Indole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00210442
CAS Number: 1670-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR