Indole-5-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-5-carbonitrile, 5-Cyanoindole, 5-Cyano-1H-indole

A235316-25G A235316-5G A235316-100G A235316-10G A235316-1G A235316-500G
103850-768EA 14 USD
103850-768 77089-774 77089-776 77261-608 77261-610 77261-612 77261-614
Indole-5-carbonitrile 97%
Indole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.16 g/mol
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 106…108 °C
Flash Pt: 121.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00005669
CAS Number: 15861-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR