Indole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A353859-1G A353859-25G A353859-5G A353859-100G A353859-10G
103826-110EA 9 USD
103826-110 103837-114 76840-230 76840-232 76840-234 77278-196 77278-198
Indole-4-carboxylic acid 98%
Indole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 213…214 °C
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00009739
CAS Number: 2124-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR