3-(Hydroxymethyl)indole 98%

Supplier: Ambeed
Warning

Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

A135866-250MG A135866-25G A135866-5G A135866-100G A135866-500G
103802-230EA 9 USD
103802-230 76688-804 76688-806 77315-102 77599-974 77624-136
3-(Hydroxymethyl)indole 98%
3-(Hydroxymethyl)indole

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 360.6 °C (760 mm Hg)
Melting Pt: 96…99 °C
Storage Temperature: Freezer
MDL Number: MFCD00005632
CAS Number: 700-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR