Indole-2-acetic acid 95%

Supplier: Ambeed
Warning

A362283-1G A362283-5G A362283-100MG A362283-250MG
103826-312EA 154 USD
103826-312 103837-326 77306-734 77306-736 77879-779 77879-780
Indole-2-acetic acid 95%
Indole-2-acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Boiling Pt: 415 °C (760 mm Hg)
Flash Pt: 204.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00957069
CAS Number: 32588-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR