Indium(III) trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Indium(III) trifluoromethanesulphonate, Indium triflate
A109212-10G
A109212-250MG
A109212-25G
A109212-1G
A109212-5G
103797-468EA
18
USD
103797-468
103823-260
76796-728
76796-730
76796-732
77310-080
77310-082
Indium(III) trifluoromethanesulfonate 98%
Indium(III) trifluoromethanesulphonate
Precaución: for Research Use Only
Formula:
C₃F₉InO₉S₃ MW: 562.03 g/mol Boiling Pt: 162 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00144478 CAS Number: 128008-30-0 UN: 3260 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...