1H-Indazole-3-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A110594-100G
A110594-10G
A110594-25G
A110594-500G
A110594-5G
103823-496EA
6
USD
103823-496
76687-528
76687-530
76687-532
76687-534
77284-024
1H-Indazole-3-carboxylic acid 98%
1H-Indazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ Boiling Pt: 443.7 °C (760 mm Hg) Melting Pt: 266…268 °C Flash Pt: 222.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00211066 CAS Number: 4498-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...