Imidazole-4-carboxaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1H-Imidazole-5-carboxaldehyde, 4-Formylimidazole, Imidazole-4-carbaldehyde
A316309-1G
A316309-25G
A316309-5G
A316309-100G
A316309-10G
A316309-500G
103825-096EA
5
USD
103825-096
103836-022
76838-862
76838-864
76838-866
77002-780
77002-782
77672-618
Imidazole-4-carboxaldehyde 98%
Imidazole-4-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₄H₄N₂O MW: 96.09 g/mol Boiling Pt: 367.8 °C (760 mm Hg) Melting Pt: 174…177 °C Flash Pt: 179.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173726 CAS Number: 3034-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...