2-Formylimidazole 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Imidazole-2-carbaldehyde, Imidazole-2-carboxaldehyde

A110938-10G A110938-25G A110938-500G A110938-1G A110938-5G A110938-100G
103797-760EA 6 USD
103797-760 103823-568 76809-940 76809-942 76809-944 77284-070 77284-072 77549-146
2-Formylimidazole 95%
2-Formylimidazole

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Melting Pt: 209 °C
Flash Pt: 137.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003544
CAS Number: 10111-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR