Imidazo[1,5-a]pyridine-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A324200-100MG A324200-250MG
103825-364EA 77 USD
103825-364 77879-671 77879-672
Imidazo[1,5-a]pyridine-1-carboxylic acid 98%
Imidazo[1,5-a]pyridine-1-carboxylic acid

Precaución: Sealed in dry, at 2…8°C

Formula: C₈H₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11846503
CAS Number: 138891-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR