Imidazo[1,2-b]pyridazine 95%

Supplier: Ambeed
Warning

A111392-100G A111392-1G A111392-5G
103797-826EA 7 USD
103797-826 103823-642 77107-960 77310-450 77310-452
Imidazo[1,2-b]pyridazine 95%
Imidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₅N₃
MW: 119.12 g/mol
Melting Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD07782103
CAS Number: 766-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR