Imidazo[1,2-b]pyridazin-6-ol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Hydroxyimidazo[1,2-b]pyridazine

A198481-1G A198481-5G
103817-022EA 29 USD
103817-022 103847-648 77085-676 77085-678
Imidazo[1,2-b]pyridazin-6-ol 97%
Imidazo[1,2-b]pyridazin-6-ol

Precaución: for Research Use Only

Formula: C₆H₅N₃O
MW: 135.1200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040161
CAS Number: 57470-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR