Imidazo[1,2-b]pyridazin-6-amine 97%

Supplier: Ambeed
Warning

A181962-250MG A181962-5G
103807-674EA 73 USD
103807-674 103846-044 76784-618 77301-764
Imidazo[1,2-b]pyridazin-6-amine 97%
Imidazo[1,2-b]pyridazin-6-amine

Precaución: for Research Use Only

Formula: C₆H₆N₄
MW: 134.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698052
CAS Number: 6653-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR