Imidazo[1,2-a]pyrimidine-7-carbaldehyde 95%

Supplier: Ambeed
Warning

A108349-100MG A108349-250MG A108349-1G
103797-330EA 916 USD
103797-330 76796-650 76796-652 77283-710
Imidazo[1,2-a]pyrimidine-7-carbaldehyde 95%
Imidazo[1,2-a]pyrimidine-7-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₅N₃O
MW: 147.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08235000
CAS Number: 375857-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR