Imidazo[1,2-a]pyrimidin-2-ylmethanamine dihydrochloride 97%

Supplier: Ambeed
Warning

A577857-100MG A577857-250MG
103828-844EA 105 USD
103828-844 103839-994 77103-720 77103-722
Imidazo[1,2-a]pyrimidin-2-ylmethanamine dihydrochloride 97%
Imidazo[1,2-a]pyrimidin-2-ylmethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀Cl₂N₄
Storage Temperature: Refrigerator
MDL Number: MFCD11506241
CAS Number: 1187928-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR