Imidazo[1,2-a]pyrimidin-2-ylmethanamine 95%

Supplier: Ambeed

A190284-1G A190284-100MG
103816-278EA 279 USD
103816-278 103846-880 77302-218 77683-494
Imidazo[1,2-a]pyrimidin-2-ylmethanamine 95%
Imidazo[1,2-a]pyrimidin-2-ylmethanamine

Precaución: For research use only.

Formula: C₇H₈N₄
MW: 148.1700 g/mol
MDL Number: MFCD06660691
CAS Number: 843609-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR