Imidazo[1,2-a]pyridine-8-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A108562-10G
A108562-250MG
A108562-25G
103797-368EA
21
USD
103797-368
103823-148
77008-806
77008-808
77008-810
Imidazo[1,2-a]pyridine-8-carboxylic acid 97%
Imidazo[1,2-a]pyridine-8-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06739251 CAS Number: 133427-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...