Imidazo[1,2-a]pyridin-7-ol 97%

Supplier: Ambeed
Warning

A174907-100MG A174907-1G A174907-250MG
103806-986EA 240 USD
103806-986 103845-286 76824-644 77082-598 77683-404
Imidazo[1,2-a]pyridin-7-ol 97%
Imidazo[1,2-a]pyridin-7-ol

Precaución: For research use only.

Formula: C₇H₆N₂O
MW: 134.135 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176749
CAS Number: 896139-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR