Imidazo[1,2-a]pyridin-6-ol 95%

Supplier: Ambeed
Warning

A154098-250MG A154098-5G A154098-100MG
103804-606EA 441 USD
103804-606 103842-786 77010-934 77300-040 77611-464
Imidazo[1,2-a]pyridin-6-ol 95%
Imidazo[1,2-a]pyridin-6-ol

Precaución: for Research Use Only

Formula: C₇H₆N₂O
MW: 134.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06739223
CAS Number: 885275-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR