5-Aminoimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: Imidazo[1,2-a]pyridin-5-amine

A203113-10G A203113-250MG A203113-1G A203113-25G A203113-5G
103817-436EA 15 USD
103817-436 103848-090 76800-424 76800-426 77086-210 77086-212 77086-214
5-Aminoimidazo[1,2-a]pyridine 98%
5-Aminoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.1506 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD05863377
CAS Number: 66358-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR