Imidazo[1,2-a]pyrazine-6-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A128970-100MG
A128970-250MG
A128970-1G
A128970-5G
103801-210EA
182
USD
103801-210
77161-790
77161-792
77313-570
77313-572
Imidazo[1,2-a]pyrazine-6-carboxylic acid 95%
Imidazo[1,2-a]pyrazine-6-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₅N₃O₂ MW: 163.13 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19441125 CAS Number: 788819-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...