Imidazo[1,2-a]pyrazine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A297640-100MG A297640-250MG A297640-1G A297640-5G
103814-148EA 30 USD
103814-148 103835-290 76802-500 76802-502 77305-714 77305-716
Imidazo[1,2-a]pyrazine-2-carboxylic acid 97%
Imidazo[1,2-a]pyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08460077
CAS Number: 77112-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR